Gene Transcripts
- Gene Symbol
- Gnat3
- Entrez Gene ID
- 286924
- Gene Name
- guanine nucleotide binding protein, alpha transducing 3
- Gene Aliases
- gnat-3
- Chromosome Location
- Chr. 4: 13827764 - 13878126 on Build Rnor_6.0
- UniGene ID
- Rn.10456
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173139.1 |
NP_775162.1
|
4 |
537 |
|
GenBank mRNA |
X65747.1 |
- |
4 |
537 |
|
View Full Product Details