Gene Transcripts

Gene Symbol
Dph1
Entrez Gene ID
287523
Gene Name
diphthamide biosynthesis 1
Gene Aliases
RGD1562694
Chromosome Location
Chr. 10: 62019379 - 62032384 on Build Rnor_6.0
UniGene ID
Rn.7589
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105809.2 NP_001099279.1 7 744
GenBank mRNA BC166808.1 - 7 744
FQ228687.1 - 7 760

View Full Product Details