Gene Transcripts

Gene Symbol
Ephb3
Entrez Gene ID
287989
Gene Name
Eph receptor B3
Gene Aliases
Chromosome Location
Chr. 11: 83527962 - 83546802 on Build Rnor_6.0
UniGene ID
Rn.131133
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105868.1 NP_001099338.1 3 718
XM_006248561.2 XP_006248623.1 3 846

View Full Product Details