Gene Transcripts

Gene Symbol
Cfhr1
Entrez Gene ID
289057
Gene Name
complement factor H-related 1
Gene Aliases
Cfhl1
Chromosome Location
Chr. 13: 56862414 - 56890208 on Build Rnor_6.0
UniGene ID
Rn.203038
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044227.1 NP_001037692.1 2 154
XM_006249941.1 XP_006250003.1 3 265
GenBank mRNA BC088173.1 - 2 154
FQ218705.1 - 2 172
FQ219461.1 - 2 175

View Full Product Details