Gene Transcripts

Gene Symbol
Ogdhl
Entrez Gene ID
290566
Gene Name
oxoglutarate dehydrogenase-like
Gene Aliases
Chromosome Location
Chr. 16: 8497494 - 8523552 on Build Rnor_6.0
UniGene ID
Rn.38202
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106062.1 NP_001099532.1 21 2913
XM_006252584.2 XP_006252646.1 3051

View Full Product Details