Gene Transcripts

Gene Symbol
Dusp9
Entrez Gene ID
293847
Gene Name
dual specificity phosphatase 9
Gene Aliases
Chromosome Location
Chr. X: 157168143 - 157172068 on Build Rnor_6.0
UniGene ID
Rn.100548
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037973.1 NP_001033062.1 4 1298
GenBank mRNA BC110044.1 - 4 1298

View Full Product Details