Gene Transcripts

Gene Symbol
Capsl
Entrez Gene ID
294795
Gene Name
calcyphosine-like
Gene Aliases
RGD1308776
Chromosome Location
Chr. 2: 58724855 - 58756261 on Build Rnor_6.0
UniGene ID
Rn.18671
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106417.1 NP_001099887.1 4 577
XM_008760763.1 XP_008758985.1 4 553

View Full Product Details