Gene Transcripts

Gene Symbol
Myd88
Entrez Gene ID
301059
Gene Name
myeloid differentiation primary response 88
Gene Aliases
Chromosome Location
Chr. 8: 128022512 - 128027461 on Build Rnor_6.0
UniGene ID
Rn.37341
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198130.1 NP_937763.1 4 763
XM_006244087.2 XP_006244149.1 5 1110
GenBank mRNA AY191270.1 - 4 763
BC097266.1 - 4 703 7459961

View Full Product Details