Gene Transcripts

Gene Symbol
Klf12
Entrez Gene ID
306110
Gene Name
Kruppel-like factor 12
Gene Aliases
Chromosome Location
Chr. 15: 84316986 - 84748539 on Build Rnor_6.0
UniGene ID
Rn.61602
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107281.1 NP_001100751.1 6, 7 1079
XM_006252408.2 XP_006252470.1 6, 7 1030
XM_006252409.2 XP_006252471.1 6, 7 1025
XM_006252410.2 XP_006252472.1 6, 7 1096
XM_006252413.2 XP_006252475.1 5, 6 878
XM_006252415.2 XP_006252477.1 5, 6 875
XM_008770870.1 XP_008769092.1 5, 6 2832
XM_008770871.1 XP_008769093.1 7, 8 1042

View Full Product Details