Gene Transcripts

Gene Symbol
Zfp451
Entrez Gene ID
316312
Gene Name
zinc finger protein 451
Gene Aliases
Chromosome Location
Chr. 9: 38396343 - 38451585 on Build Rnor_6.0
UniGene ID
Rn.163059
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033705.1 NP_001028877.1 5 507
GenBank mRNA BC101930.1 - 5 507

View Full Product Details