Gene Transcripts

Gene Symbol
PABPC4
Entrez Gene ID
8761
Gene Name
poly(A) binding protein cytoplasmic 4
Gene Aliases
APP-1, APP1, PABP4, iPABP
Chromosome Location
Chr. 1: 39560813 - 39576849 on Build GRCh38
UniGene ID
Hs.169900
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135653.1 NP_001129125.1 5 1569
NM_001135654.1 NP_001129126.1 5 1569
NM_003819.3 NP_003810.1 5 1569
GenBank mRNA BC029969.1 - 5 1121
BC065540.1 - 5 1472 6061561
BC071591.1 - 5 1507 5265829
BC094755.1 - 5 1510 30331896
D12799.1 - 2 104
U33818.1 - 5 824
U75686.1 - 5 1465

View Full Product Details