Gene Transcripts

Gene Symbol
Ankrd40
Entrez Gene ID
690586
Gene Name
ankyrin repeat domain 40
Gene Aliases
Chromosome Location
Chr. 10: 82032656 - 82045937 on Build Rnor_6.0
UniGene ID
Rn.17667
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134699.1 NP_001128171.1 2 449
GenBank mRNA BC166790.1 - 2 449

View Full Product Details