Gene Transcripts

Gene Symbol
GOLGA5
Entrez Gene ID
9950
Gene Name
golgin A5
Gene Aliases
GOLIM5, RFG5, ret-II
Chromosome Location
Chr. 14: 92794231 - 92839963 on Build GRCh38
UniGene ID
Hs.104320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005113.3 NP_005104.3 4, 5 1255
XM_011537420.2 XP_011535722.1 4, 5 1255
GenBank mRNA AF085199.1 - 4, 5 1130
AJ132949.1 - 3, 4 999
AK293077.1 - 4, 5 1158
AK303789.1 - 5, 6 888
AY644768.1 - 3, 4 1035
BC023021.1 - 4, 5 1163 4999710

View Full Product Details