Gene Transcripts

Gene Symbol
MISP3
Entrez Gene ID
113230
Gene Name
MISP family member 3
Gene Aliases
Chromosome Location
Chr. 19: 14073009 - 14075062 on Build GRCh38
UniGene ID
Hs.372775
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291291.1 NP_001278220.1 3 1214
NR_111941.1 3 1279
GenBank mRNA BC011002.2 - 3 1214

View Full Product Details