Gene Transcripts

Gene Symbol
MOK
Entrez Gene ID
5891
Gene Name
MOK protein kinase
Gene Aliases
RAGE, RAGE-1, RAGE1, STK30
Chromosome Location
Chr. 14: 102224450 - 102305200 on Build GRCh38
UniGene ID
Hs.104119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272011.1 NP_001258940.1 4 495
NM_014226.2 NP_055041.1 5 585
XM_006720225.2 XP_006720288.1 5 585
XM_006720226.2 XP_006720289.1 5 585
XM_011537052.1 XP_011535354.1 5 585
XM_017021549.1 XP_016877038.1 5 585
XM_017021550.1 XP_016877039.1 4 484
XM_017021551.1 XP_016877040.1 6 525
XM_017021552.1 XP_016877041.1 5 626
XM_017021553.1 XP_016877042.1 3 423
XM_017021554.1 XP_016877043.1 3 419
XM_017021555.1 XP_016877044.1 5 585
XM_017021556.1 XP_016877045.1 5 585
XM_017021557.1 XP_016877046.1 4 485
XM_017021558.1 XP_016877047.1 4 451
XM_017021559.1 XP_016877048.1 3 410
GenBank mRNA AB022694.1 - 5 579
AK094747.1 - 3 392
AK302349.1 - 4 480
AK312778.1 - 5 576
BC043179.1 - 2 1177 5287879
BC053536.1 - 5 561 5175851

View Full Product Details