Gene Transcripts

Gene Symbol
Ifne
Entrez Gene ID
230405
Gene Name
interferon epsilon
Gene Aliases
Ifn-tau-1, Ifne1, Ifnt1, Infe1
Chromosome Location
Chr. 4: 88879538 - 88880201 on Build GRCm38
UniGene ID
Mm.246593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177348.2 NP_796322.1 1 483
GenBank mRNA AY190044.1 - 1 483
BC104376.2 - 1 482 40050053
BC104377.2 - 1 482 40050054

View Full Product Details