Gene Transcripts

Gene Symbol
Mlana
Entrez Gene ID
77836
Gene Name
melan-A
Gene Aliases
A930034P04Rik, Mart1
Chromosome Location
Chr. 19: 29697548 - 29708447 on Build GRCm38
UniGene ID
Mm.138437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029993.1 NP_084269.1 5 403
GenBank mRNA AK020928.1 - 5 422
BC111114.1 - 5 403 40053172

View Full Product Details