Gene Transcripts

Gene Symbol
BSG
Entrez Gene ID
682
Gene Name
basigin (Ok blood group)
Gene Aliases
5F7, CD147, EMMPRIN, OK, TCSF
Chromosome Location
Chr. 19: 571277 - 583493 on Build GRCh38
UniGene ID
Hs.501293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322243.1 NP_001309172.1 8 1103
NM_001728.3 NP_001719.2 9 1455
NM_198589.2 NP_940991.1 8 1107
NM_198590.2 NP_940992.1 8 1162
NM_198591.2 NP_940993.1 7 1005
XM_017027173.1 XP_016882662.1 9 1340
GenBank mRNA AF548371.1 - 9 1334
AK095912.1 - 7 1523
AK296858.1 - 6 620
AK297880.1 - 9 1313
AK302231.1 - 8 975
AY358113.1 - 9 1326
BC009040.2 - 8 970 3867352
D45131.1 - 8 955
GU557064.1 - 8 1129
GU557065.1 - 7 972
L10240.1 - 8 940
L20471.1 - 8 971
M87879.1 - 8 948
X64364.1 - 8 983

View Full Product Details