Gene Transcripts

Gene Symbol
Trim15
Entrez Gene ID
69097
Gene Name
tripartite motif-containing 15
Gene Aliases
1810012B10Rik
Chromosome Location
Chr. 17: 36860690 - 36868085 on Build GRCm38
UniGene ID
Mm.307027
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024134.2 NP_001019305.1 2 496
NM_001177872.1 NP_001171343.1 2 519
GenBank mRNA AK007445.1 - 2 520
BC027186.1 - 2 496 4215801

View Full Product Details