Gene Transcripts

Gene Symbol
Asb18
Entrez Gene ID
316614
Gene Name
ankyrin repeat and SOCS box-containing 18
Gene Aliases
Chromosome Location
Chr. 9: 97085234 - 97151832 on Build Rnor_6.0
UniGene ID
Rn.45037
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108231.1 NP_001101701.1 1, 2 199

View Full Product Details