Gene Transcripts

Gene Symbol
Armcx6
Entrez Gene ID
363496
Gene Name
armadillo repeat containing, X-linked 6
Gene Aliases
Chromosome Location
Chr. X: 105565733 - 105573353 on Build Rnor_6.0
UniGene ID
Rn.225307
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007757.1 NP_001007758.1 3 790
XM_006257228.2 XP_006257290.1 4 911
XM_008773423.1 XP_008771645.1 7 2206
XM_008773424.1 XP_008771646.1 7 3208
XM_008773425.1 XP_008771647.1 4 935
GenBank mRNA BC081893.1 - 3 790

View Full Product Details