Gene Transcripts
- Gene Symbol
- Rpe65
- Entrez Gene ID
- 89826
- Gene Name
- retinal pigment epithelium 65
- Gene Aliases
- Chromosome Location
- Chr. 2: 266141581 - 266169197 on Build Rnor_6.0
- UniGene ID
- Rn.76724
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053562.3 |
NP_446014.2
|
11 |
1240 |
|
GenBank mRNA |
AF035673.1 |
- |
11 |
1242 |
|
View Full Product Details