Gene Transcripts

Gene Symbol
ECD
Entrez Gene ID
11319
Gene Name
ecdysoneless cell cycle regulator
Gene Aliases
GCR2, HSGT1, SGT1
Chromosome Location
Chr. 10: 73134524 - 73168095 on Build GRCh38
UniGene ID
Hs.631822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135752.1 NP_001129224.1 15 2093
NM_001135753.1 NP_001129225.1 13 1865
NM_007265.2 NP_009196.1 14 1994
NR_024203.1 12 1583
GenBank mRNA AK022616.1 - 14 1942
AK225519.1 - 13 1841
AK315711.1 - 15 2066
BC000721.2 - 14 1958 3504956
CR457407.1 - 13 1751
D88208.1 - 14 1958

View Full Product Details