Gene Transcripts

Gene Symbol
ZFAT
Entrez Gene ID
57623
Gene Name
zinc finger and AT-hook domain containing
Gene Aliases
AITD3, ZFAT1, ZNF406
Chromosome Location
Chr. 8: 134477788 - 134713049 on Build GRCh38
UniGene ID
Hs.446172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029939.3 NP_001025110.2 7 2424
NM_001167583.2 NP_001161055.1 6 2265
NM_001174157.1 NP_001167628.1 5 2156
NM_001174158.1 NP_001167629.1 6 2265
NM_001289394.1 NP_001276323.1 7 2501
NM_020863.3 NP_065914.2 6 2342
NR_110323.1 6 2342
XM_011517203.1 XP_011515505.1 6 2239
XM_011517204.2 XP_011515506.1 5 2062
XM_011517206.1 XP_011515508.1 6 2263
XM_017013716.1 XP_016869205.1 5 2137
GenBank mRNA AB040918.1 - 4 1740
AB167738.1 - 6 2331
AB167739.1 - 7 2490
AB167740.1 - 7 2417
AB167741.1 - 6 2258
AK300133.1 - 7 1951
AK300324.1 - 6 2261
AK300471.1 - 6 2327
AL832066.1 - 6 2342
BC012989.2 - 6 2256
BC025423.2 - 4 1934
BC039059.1 - 1 26
BC098405.1 - 6 2265
BC101766.1 - 6 2255 8069272
BC101768.1 - 6 2255 8069274
BC143519.1 - 5 2070 9052028

View Full Product Details