Gene Transcripts

Gene Symbol
ACIN1
Entrez Gene ID
22985
Gene Name
apoptotic chromatin condensation inducer 1
Gene Aliases
ACINUS, ACN, fSAP152
Chromosome Location
Chr. 14: 23058564 - 23095614 on Build GRCh38
UniGene ID
Hs.124490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164814.1 NP_001158286.1 7 2487
NM_001164815.1 NP_001158287.1 6 2367
NM_014977.3 NP_055792.1 7 2487
XM_005267415.3 XP_005267472.1 7 2291
XM_005267416.3 XP_005267473.1 7 2291
XM_006720081.3 XP_006720144.1 7 2291
XM_011536570.2 XP_011534872.1 7 2291
GenBank mRNA AB014570.2 - 7 1996
AF124726.1 - 7 2486
AK299034.1 - 6 1878
AL833228.1 - 2 340
BC140805.1 - 7 2256 9021673

View Full Product Details