Gene Transcripts

Gene Symbol
C9orf24
Entrez Gene ID
84688
Gene Name
chromosome 9 open reading frame 24
Gene Aliases
CBE1, NYD-SP22, SMRP1, bA573M23.4
Chromosome Location
Chr. 9: 34379019 - 34400833 on Build GRCh38
UniGene ID
Hs.50334
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252195.1 NP_001239124.1 2, 3 160
NM_032596.3 NP_115985.2 4, 5 699
NM_147168.1 NP_671697.1 1, 2 241
NM_147169.2 NP_671698.1 1, 2 241
XM_005251616.4 XP_005251673.1 4, 5 3681
XM_005251618.4 XP_005251675.1 2, 3 228
XM_011518055.2 XP_011516357.1 2, 3 502
GenBank mRNA AF367474.1 - 1, 2 180
AK058087.1 - 4, 5 637
AK094751.1 - 1, 2 230
AK225981.1 - 1, 2 159
AW961020.1 - 1, 2 241
BC022084.1 - 4, 5 684 4825995
BC029484.1 - 4, 5 699 5271301
BI561111.1 - 2, 3 125
CB854584.1 - 1, 2 210

View Full Product Details