Gene Transcripts

Gene Symbol
SPATA31A5
Entrez Gene ID
727905
Gene Name
SPATA31 subfamily A member 5
Gene Aliases
FAM75A5
Chromosome Location
Chr. 9: 60914372 - 60920653 on Build GRCh38
UniGene ID
Hs.645497
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113541.1 NP_001107013.1 1 181
XM_011545671.2 XP_011543973.1 1 216

Gene Symbol
SPATA31A1
Entrez Gene ID
647060
Gene Name
SPATA31 subfamily A member 1
Gene Aliases
C9orf36, C9orf36A, FAM75A1, FAM75A2, SPATA31A2
Chromosome Location
Chr. 9: 39355667 - 39361962 on Build GRCh38
UniGene ID
Hs.575677
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085452.2 NP_001078921.2 1 214
XM_011518006.1 XP_011516308.1 1 216
GenBank mRNA AK302569.1 - 1 214
BC146855.1 - 1 173 9056783

Gene Symbol
SPATA31A3
Entrez Gene ID
727830
Gene Name
SPATA31 subfamily A member 3
Gene Aliases
FAM75A3
Chromosome Location
Chr. 9: 66986301 - 66992550 on Build GRCh38
UniGene ID
Hs.632078
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083124.1 NP_001076593.1 1 181

Gene Symbol
SPATA31A7
Entrez Gene ID
26165
Gene Name
SPATA31 subfamily A member 7
Gene Aliases
AEP1, C9orf36A, FAM75A4, FAM75A7, SPATA31A4
Chromosome Location
Chr. 9: 61190036 - 61196280 on Build GRCh38
UniGene ID
Hs.123004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015667.2 NP_056482.2 1 181
GenBank mRNA BC040528.1 - 1 181

View Full Product Details