Gene Transcripts

Gene Symbol
DEFB103A
Entrez Gene ID
414325
Gene Name
defensin beta 103A
Gene Aliases
BD-3, DEFB-3, DEFB103, DEFB3, HBD3, HBP-3, HBP3, hBD-3
Chromosome Location
Chr. 8: 7881204 - 7882664 on Build GRCh38
UniGene ID
Hs.283082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081551.3 NP_001075020.1 1 221

Gene Symbol
DEFB103B
Entrez Gene ID
55894
Gene Name
defensin beta 103B
Gene Aliases
BD-3, DEFB-3, DEFB103, DEFB3, HBD-3, HBD3, HBP-3, HBP3
Chromosome Location
Chr. 8: 7428888 - 7430348 on Build GRCh38
UniGene ID
Hs.283082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018661.4 NP_061131.1 1 221
GenBank mRNA AB037972.1 - 1 26
AF295370.1 - 1 38
AF301470.1 - 1 221
AJ237673.1 - 1 35

View Full Product Details