Gene Transcripts

Gene Symbol
Sh2d1b1
Entrez Gene ID
26904
Gene Name
SH2 domain containing 1B1
Gene Aliases
EAT-2, EAT-2A, Eat2, Eat2a, Sh2d1b
Chromosome Location
Chr. 1: 170277324 - 170285662 on Build GRCm38
UniGene ID
Mm.57009
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012009.4 NP_036139.3 251
GenBank mRNA BC116698.1 - 251
BC117508.1 - 251 40105515

View Full Product Details