Gene Transcripts

Gene Symbol
NEPRO
Entrez Gene ID
25871
Gene Name
nucleolus and neural progenitor protein
Gene Aliases
C3orf17, NET17
Chromosome Location
Chr. 3: 113002444 - 113019733 on Build GRCh38
UniGene ID
Hs.591288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319112.1 NP_001306041.1 4, 5 442
NM_001319114.1 NP_001306043.1 4, 5 488
NM_001319115.1 NP_001306044.1 4, 5 484
NM_015412.3 NP_056227.2 5, 6 614
XM_005247276.2 XP_005247333.1 5, 6 581
XM_017006102.1 XP_016861591.1 5, 6 577
XM_017006103.1 XP_016861592.1 4, 5 472
GenBank mRNA AK024325.1 - 5, 6 572
AK295165.1 - 4, 5 373
AK301161.1 - 4, 5 402
AK304083.1 - 4, 5 417
AL117573.1 - 5, 6 556
AL537045.3 - 4, 5 409
BC014673.2 - 4, 5 1072
BC067743.1 - 5, 6 574 4812681

View Full Product Details