Gene Transcripts

Gene Symbol
LARP7
Entrez Gene ID
51574
Gene Name
La ribonucleoprotein domain family member 7
Gene Aliases
ALAZS, HDCMA18P, PIP7S
Chromosome Location
Chr. 4: 112636964 - 112657592 on Build GRCh38
UniGene ID
Hs.713663
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267039.1 NP_001253968.1 13, 14 2063
NM_015454.2 NP_056269.1 11, 12 1727
NM_016648.3 NP_057732.2 11, 12 1855
NR_049768.1 11, 12 1752
GenBank mRNA AF068284.1 - 10, 11 1004
AK000274.1 - 11, 12 1665
AK225900.1 - 8, 9 1224
AL049996.1 - 5, 6 702
BC066945.1 - 13, 14 1901 4837780
BC107709.1 - 5, 6 696
BX647100.1 - 11, 12 1589
EU667388.1 - 11, 12 1598

View Full Product Details