Gene Transcripts

Gene Symbol
CXorf57
Entrez Gene ID
55086
Gene Name
chromosome X open reading frame 57
Gene Aliases
Chromosome Location
Chr. X: 106611930 - 106679443 on Build GRCh38
UniGene ID
Hs.274267
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184782.1 NP_001171711.1 11 1962
NM_018015.5 NP_060485.4 12 2253
GenBank mRNA AK024253.1 - 11 1236
AK291730.1 - 12 2202
BC011483.1 - 12 2174 3453700
BC033709.1 - 12 2165 3910777
BC070110.1 - 12 2204 5264812

View Full Product Details