Gene Transcripts

Gene Symbol
ZNF77
Entrez Gene ID
58492
Gene Name
zinc finger protein 77
Gene Aliases
pT1
Chromosome Location
Chr. 19: 2933218 - 2944971 on Build GRCh38
UniGene ID
Hs.38004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021217.2 NP_067040.1 4 672
XM_017027081.1 XP_016882570.1 3 535
XM_017027082.1 XP_016882571.1 4 697
GenBank mRNA AK290979.1 - 4 608
BC043354.1 - 4 633 6014754
BC062561.1 - 4 634 6644579
X65230.1 - 2 264

View Full Product Details