Gene Transcripts

Gene Symbol
XYLT1
Entrez Gene ID
64131
Gene Name
xylosyltransferase 1
Gene Aliases
DBQD2, PXYLT1, XT-I, XT1, XTI, XYLTI, xylT-I
Chromosome Location
Chr. 16: 17102324 - 17470934 on Build GRCh38
UniGene ID
Hs.22907
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022166.3 NP_071449.1 9 1930
XM_017023539.1 XP_016879028.1 9 1983
XM_017023540.1 XP_016879029.1 9 1983
GenBank mRNA AJ277441.1 - 7 1449
AJ539163.1 - 9 1930

View Full Product Details