Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032856.3 | NP_116245.2 | 3 | 227 | |
NR_130944.1 | 2 | 664 | |||
NR_130945.1 | 3 | 227 | |||
NR_130946.1 | 3 | 234 | |||
NR_130947.1 | 3 | 227 | |||
GenBank mRNA | AF161382.1 | - | 3 | 158 | |
AK027794.1 | - | 3 | 164 | ||
AK055578.1 | - | 2 | 664 | ||
AK055729.1 | - | 3 | 164 | ||
AK090406.1 | - | 2 | 1360 | ||
AK225068.1 | - | 3 | 167 | ||
AK225199.1 | - | 3 | 164 | ||
AK295372.1 | - | 3 | 158 | ||
BC014115.2 | - | 3 | 165 | ||
BC020252.2 | - | 2 | 116 | ||
BC050648.1 | - | 3 | 184 | ||
BC063392.1 | - | 3 | 195 | 5168550 | |
BU601517.1 | - | 237 | |||
BX446522.2 | - | 3 | 157 | ||
CD514627.1 | - | 3 | 150 | ||
CN481562.1 | - | 3 | 163 |