Gene Transcripts

Gene Symbol
DUSP9
Entrez Gene ID
1852
Gene Name
dual specificity phosphatase 9
Gene Aliases
MKP-4, MKP4
Chromosome Location
Chr. X: 153642443 - 153651326 on Build GRCh38
UniGene ID
Hs.144879
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318503.1 NP_001305432.1 4 1199
NM_001395.3 NP_001386.1 4 1145
XM_011531123.1 XP_011529425.1 4 1058
XM_011531124.1 XP_011529426.1 4 1191
GenBank mRNA AK314250.1 - 4 1145
BC034936.1 - 4 853
BC042166.1 - 4 853
BC060837.1 - 4 1199 30339218
U87167.1 - 2 141
Y08302.1 - 4 1054

View Full Product Details