Gene Transcripts

Gene Symbol
KRTAP13-1
Entrez Gene ID
140258
Gene Name
keratin associated protein 13-1
Gene Aliases
KAP13.1, KRTAP13.1
Chromosome Location
Chr. 21: 30396074 - 30396822 on Build GRCh38
UniGene ID
Hs.407653
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181599.2 NP_853630.2 1 584
GenBank mRNA AB096939.1 - 1 584
AJ457066.1 - 1 452

View Full Product Details