Gene Transcripts

Gene Symbol
FNIP2
Entrez Gene ID
57600
Gene Name
folliculin interacting protein 2
Gene Aliases
FNIPL, MAPO1
Chromosome Location
Chr. 4: 158768995 - 158908050 on Build GRCh38
UniGene ID
Hs.652441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323916.1 NP_001310845.1 17 4280
NM_020840.2 NP_065891.1 17 4275
XM_005263156.1 XP_005263213.1 18 4365
XM_005263158.1 XP_005263215.1 18 4165
XM_011532154.2 XP_011530456.1 18 5145
XM_017008487.1 XP_016863976.1 19 4314
GenBank mRNA AB040883.1 - 17 4132
AK022968.1 - 5 2072
BC047069.1 - 1 115

View Full Product Details