Gene Transcripts
- Gene Symbol
- ZNF433
- Entrez Gene ID
- 163059
- Gene Name
- zinc finger protein 433
- Gene Aliases
- Chromosome Location
- Chr. 19: 12014717 - 12035741 on Build GRCh38
- UniGene ID
- Hs.667776
- Species
- Homo sapiens
- Gene Symbol
- ZNF878
- Entrez Gene ID
- 729747
- Gene Name
- zinc finger protein 878
- Gene Aliases
- Chromosome Location
- Chr. 19: 12043647 - 12052967 on Build GRCh38
- UniGene ID
- Hs.704229
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DA007842.1 |
- |
5 |
531 |
|
View Full Product Details