Gene Transcripts

Gene Symbol
PLAC9
Entrez Gene ID
219348
Gene Name
placenta specific 9
Gene Aliases
Chromosome Location
Chr. 10: 80132502 - 80145028 on Build GRCh38
UniGene ID
Hs.204947
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012973.1 NP_001012991.1 4 651
GenBank mRNA BC066348.1 - 4 651
BC090922.1 - 4 651 30331098

View Full Product Details