Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175735.3 | NP_783862.2 | 6 | 660 | |
XM_017003751.1 | XP_016859240.1 | 7 | 1825 | ||
GenBank mRNA | BC100882.3 | - | 5 | 568 | |
BC100883.2 | - | 6 | 629 | 40003266 | |
BC100884.1 | - | 5 | 568 | 40003273 | |
BC100885.2 | - | 5 | 568 | 40003274 |