Gene Transcripts

Gene Symbol
NKAIN3
Entrez Gene ID
286183
Gene Name
Na+/K+ transporting ATPase interacting 3
Gene Aliases
FAM77D
Chromosome Location
Chr. 8: 62248942 - 62999652 on Build GRCh38
UniGene ID
Hs.654662
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304533.1 NP_001291462.1 2 308
NR_130764.1 2 308
XM_011517511.2 XP_011515813.1 2 308
XM_011517512.2 XP_011515814.1 2 308
XM_017013359.1 XP_016868848.1 2 308
XM_017013360.1 XP_016868849.1 2 308
XM_017013361.1 XP_016868850.1 2 1690
GenBank mRNA AK096949.1 - 2 308
AK299689.1 - 2 581

View Full Product Details