Gene Transcripts

Gene Symbol
MS4A10
Entrez Gene ID
341116
Gene Name
membrane spanning 4-domains A10
Gene Aliases
CD20L7, MS4A9
Chromosome Location
Chr. 11: 60785332 - 60801305 on Build GRCh38
UniGene ID
Hs.591956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206893.3 NP_996776.2 5 517
XM_011544989.1 XP_011543291.1 5 517
XM_011544991.2 XP_011543293.1 4 328
GenBank mRNA AK122633.1 - 5 518
BC137259.1 - 5 467 9021256
BC137263.1 - 5 467 9021260

View Full Product Details