Gene Transcripts

Gene Symbol
SKA2
Entrez Gene ID
348235
Gene Name
spindle and kinetochore associated complex subunit 2
Gene Aliases
FAM33A
Chromosome Location
Chr. 17: 59109947 - 59155439 on Build GRCh38
UniGene ID
Hs.463607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100595.1 NP_001094065.1 3 587
NM_182620.3 NP_872426.1 4 589
XM_011524745.2 XP_011523047.1 4 423
GenBank mRNA AK022820.1 - 3 144
AK056473.1 - 4 341
BC017873.1 - 4 350 4685972
BC106003.1 - 4 314 6143473
DA133216.1 - 3 325
DB158057.1 - 2 325

View Full Product Details