Gene Transcripts

Gene Symbol
Dmwd
Entrez Gene ID
13401
Gene Name
dystrophia myotonica-containing WD repeat motif
Gene Aliases
DMR-N9, Dm9
Chromosome Location
Chr. 7: 19076089 - 19082776 on Build GRCm38
UniGene ID
Mm.2852
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010058.2 NP_034188.2 2 671
GenBank mRNA AK086899.1 - 1 231
BC089027.1 - 2 903 30615314
CF169673.1 - 2 334
S60312.1 - 2 311

View Full Product Details