Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008585.2 | NP_032611.2 | 2, 3 | 144 | |
GenBank mRNA | AK093914.1 | - | 2, 3 | 105 | |
AK143906.1 | - | 2, 3 | 101 | ||
AK164734.1 | - | 2, 3 | 99 | ||
BC015258.1 | - | 2, 3 | 144 | 4241252 | |
M74897.1 | - | 2, 3 | 143 |