Gene Transcripts

Gene Symbol
Mns1
Entrez Gene ID
17427
Gene Name
meiosis-specific nuclear structural protein 1
Gene Aliases
AW546487
Chromosome Location
Chr. 9: 72438518 - 72458576 on Build GRCm38
UniGene ID
Mm.387671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008613.3 NP_032639.1 2 229
GenBank mRNA AK013048.1 - 2 196
AK132772.1 - 2 204
BC046624.1 - 2 142 6314904
D14849.1 - 2 260

View Full Product Details