Gene Transcripts

Gene Symbol
Chst4
Entrez Gene ID
26887
Gene Name
carbohydrate (chondroitin 6/keratan) sulfotransferase 4
Gene Aliases
GST-3, Gn6st-2, HEC-GlcNAc6ST
Chromosome Location
Chr. 8: 110029075 - 110039375 on Build GRCm38
UniGene ID
Mm.89207
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011998.4 NP_036128.3 2 787
GenBank mRNA AF109155.1 - 2 1011
AK009113.1 - 2 788
BC057886.1 - 2 807 4987327

View Full Product Details