Gene Transcripts

Gene Symbol
Larp7
Entrez Gene ID
28036
Gene Name
La ribonucleoprotein domain family, member 7
Gene Aliases
C330027G06Rik, D3Wsu161e
Chromosome Location
Chr. 3: 127536714 - 127553349 on Build GRCm38
UniGene ID
Mm.291032
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138593.2 NP_613059.2 12 1744
GenBank mRNA AK012726.1 - 1 46
BC029178.1 - 4 417
BC132266.1 - 4 415
BC132268.1 - 4 415
BM946683.1 - 6 612

View Full Product Details