Gene Transcripts

Gene Symbol
Mga
Entrez Gene ID
29808
Gene Name
MAX gene associated
Gene Aliases
AV312082, C130042M01Rik, C80739, D030062C11Rik, Mad5
Chromosome Location
Chr. 2: 119871646 - 119970129 on Build GRCm38
UniGene ID
Mm.437053
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013720.2 NP_038748.2 10, 11 3826
GenBank mRNA AF205935.1 - 9, 10 3643
CD539587.1 - 2, 3 425

View Full Product Details